Key Documents
348392
Sigma-Aldrich
2,3,5-Trifluorophenol
98%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
F3C6H2OH
CAS Number:
Molecular Weight:
148.08
MDL number:
UNSPSC Code:
12352103
assay
98%
refractive index
n20/D 1.462 (lit.)
bp
57 °C/29 mmHg (lit.)
mp
29 °C (lit.)
density
1.457 g/mL at 25 °C (lit.)
SMILES string
Oc1cc(F)cc(F)c1F
pictograms
FlameExclamation mark
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
4.1B - Flammable solid hazardous materials
wgk_germany
WGK 3
flash_point_f
129.2 °F - closed cup
flash_point_c
54 °C - closed cup
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)