Key Documents
348066
Sigma-Aldrich
[1,2-Bis(diphenylphosphino)ethane]tetracarbonyltungsten(0)
97%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
[(C6H5)2PCH2CH2P(C6H5)2]W(CO)4
CAS Number:
Molecular Weight:
694.30
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay
97%
mp
206-208 °C (lit.)
SMILES string
[W].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].C(CP(c1ccccc1)c2ccccc2)P(c3ccccc3)c4ccccc4
Choose from one of the most recent versions:
Certificates of Analysis (COA)