Key Documents
323926
Sigma-Aldrich
4-Amino-2,6-dibromophenol
96%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
H2NC6H2(Br)2OH
CAS Number:
Molecular Weight:
266.92
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
96%
mp
195-196 °C (lit.)
SMILES string
Nc1cc(Br)c(O)c(Br)c1
pictograms
Exclamation mark
signalword
Warning
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)