Key Documents
323845
Sigma-Aldrich
(−)-TAPA
Synonym(s):
(−)-2-(2,4,5,7-Tetranitro-9-fluorenylideneaminooxy)propionic acid
Sign Into View Organizational & Contract Pricing
Select a Size
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C16H9N5O11
CAS Number:
Molecular Weight:
447.27
MDL number:
UNSPSC Code:
12352103
optical activity
[α]20/D −91°, c = 1.6 in dioxane
impurities
<2% acetic acid
mp
196-199 °C (lit.)
SMILES string
C[C@@H](O\N=C1\c2cc(cc(c2-c3c1cc(cc3[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O)C(O)=O
Choose from one of the most recent versions:
Certificates of Analysis (COA)