Key Documents
316423
Sigma-Aldrich
5-Isopropyl-3-methylphenol
98%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
(CH3)2CHC6H3(CH3)OH
CAS Number:
Molecular Weight:
150.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
98%
mp
49-51 °C (lit.)
SMILES string
CC(C)c1cc(C)cc(O)c1
Choose from one of the most recent versions:
Certificates of Analysis (COA)