Key Documents
307211
Sigma-Aldrich
Chlorodibromofluoromethane
technical grade, 90%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
ClCBr2F
CAS Number:
Molecular Weight:
226.27
MDL number:
UNSPSC Code:
12352103
grade
technical grade
assay
90%
refractive index
n20/D 1.4772 (lit.)
bp
79-80 °C (lit.)
density
2.317 g/mL at 25 °C (lit.)
SMILES string
FC(Cl)(Br)Br
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)