Key Documents
293032
Sigma-Aldrich
Stearyltributylphosphonium bromide
95%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
CH3(CH2)17P[(CH2)3CH3]3Br
CAS Number:
Molecular Weight:
537.72
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
assay
95%
mp
70-72 °C (lit.)
SMILES string
[Br-].CCCCCCCCCCCCCCCCCC[P+](CCCC)(CCCC)CCCC
Choose from one of the most recent versions:
Certificates of Analysis (COA)