Key Documents
283568
Sigma-Aldrich
4-Ethoxybenzonitrile
99%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
C2H5OC6H4CN
CAS Number:
Molecular Weight:
147.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
99%
bp
258 °C (lit.)
mp
62-64 °C (lit.)
SMILES string
CCOc1ccc(cc1)C#N
Choose from one of the most recent versions:
Certificates of Analysis (COA)