Key Documents
283185
Sigma-Aldrich
α,α-Dichloro-4-fluorotoluene
97%
Synonym(s):
1-(Dichloromethyl)-4-fluorobenzene, 4-Fluorobenzal chloride
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
FC6H4CHCl2
CAS Number:
Molecular Weight:
179.02
EC Number:
MDL number:
UNSPSC Code:
12352103
assay
97%
refractive index
n20/D 1.528 (lit.)
bp
81-83 °C/15 mmHg (lit.)
density
1.332 g/mL at 25 °C (lit.)
SMILES string
Fc1ccc(cc1)C(Cl)Cl
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)