Key Documents
281271
Sigma-Aldrich
1,4,7,10,13,16-Hexathiacyclooctadecane
Synonym(s):
18-Ane-S6
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C12H24S6
CAS Number:
Molecular Weight:
360.71
Beilstein/REAXYS Number:
143957
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
mp
92-94 °C (lit.)
SMILES string
C1CSCCSCCSCCSCCSCCS1
InChI
1S/C12H24S6/c1-2-14-5-6-16-9-10-18-12-11-17-8-7-15-4-3-13-1/h1-12H2
InChI key
HELCSRZMDRYPIJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
pictograms
Exclamation mark
signalword
Warning
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)