All Photos(1)
Documents
280615
Sigma-Aldrich
N-(2-Hydroxyethyl)salicylamide
98%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
2-(HO)C6H4CONHCH2CH2OH
CAS Number:
Molecular Weight:
181.19
EC Number:
MDL number:
UNSPSC Code:
12352103
assay
98%
bp
210-215 °C/2 mmHg (lit.)
mp
115-117 °C (lit.)
SMILES string
OCCNC(=O)c1ccccc1O
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.