Key Documents
275085
Sigma-Aldrich
N,N′-Bis(salicylidene)-1,6-hexanediamine
99%
Synonym(s):
N,N′-Hexamethylenebis(salicylimine)
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
HOC6H4CH=N(CH2)6N=CHC6H4OH
CAS Number:
Molecular Weight:
324.42
EC Number:
MDL number:
UNSPSC Code:
12171500
assay
99%
mp
68-69 °C (lit.)
SMILES string
Oc1ccccc1\C=N\CCCCCC\N=C\c2ccccc2O
Choose from one of the most recent versions:
Certificates of Analysis (COA)