Key Documents
249041
Sigma-Aldrich
cis-11-Tetradecen-1-ol
95%
Synonym(s):
(Z)-11-TDOL, Z-11-TDOL
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
C2H5CH=CH(CH2)10OH
CAS Number:
Molecular Weight:
212.37
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
95%
refractive index
n20/D 1.457 (lit.)
SMILES string
[H]\C(CC)=C(/[H])CCCCCCCCCCO
Choose from one of the most recent versions:
Certificates of Analysis (COA)