Key Documents
246662
Sigma-Aldrich
3′-Amino-4′-methoxyacetanilide
technical grade, 90%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
CH3CONHC6H3(NH2)OCH3
CAS Number:
Molecular Weight:
180.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade
technical grade
assay
90%
SMILES string
COc1ccc(NC(C)=O)cc1N
Choose from one of the most recent versions:
Certificates of Analysis (COA)