Key Documents
244236
Sigma-Aldrich
1-(2,4,6-Triisopropylbenzenesulfonyl)imidazole
99%
Synonym(s):
1-(2,4,6-Triisopropylphenylsulfonyl)imidazole, TPSI
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C18H26N2O2S
CAS Number:
Molecular Weight:
334.48
Beilstein/REAXYS Number:
962743
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
99%
mp
118-120 °C (lit.)
SMILES string
CC(C)c1cc(C(C)C)c(c(c1)C(C)C)S(=O)(=O)n2ccnc2
InChI
1S/C18H26N2O2S/c1-12(2)15-9-16(13(3)4)18(17(10-15)14(5)6)23(21,22)20-8-7-19-11-20/h7-14H,1-6H3
InChI key
AGGRGODMKWLSDE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Coupling or condensing agent in oligonucleotide synthesis.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)