Key Documents
235873
Sigma-Aldrich
Folic acid dihydrate
97%
Synonym(s):
PGA, Pteroylglutamic acid
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C19H19N7O6 · 2H2O
CAS Number:
Molecular Weight:
477.43
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
assay
97%
optical activity
[α]20/D +18°, c = 0.5 in 0.1 M NaOH
SMILES string
O.O.NC1=NC(=O)c2nc(CNc3ccc(cc3)C(=O)N[C@@H](CCC(O)=O)C(O)=O)cnc2N1
Choose from one of the most recent versions:
Certificates of Analysis (COA)