Key Documents
225843
Sigma-Aldrich
2-[2-(2-Methoxyethoxy)ethoxy]-1,3-dioxolane
97%
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C8H16O5
CAS Number:
Molecular Weight:
192.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
97%
refractive index
n20/D 1.439 (lit.)
bp
96 °C/0.3 mmHg (lit.)
density
1.102 g/mL at 25 °C (lit.)
SMILES string
COCCOCCOC1OCCO1
Choose from one of the most recent versions:
Certificates of Analysis (COA)