Key Documents
222291
Sigma-Aldrich
3-Methylthio-1,2-propanediol
99%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
CH3SCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
122.19
EC Number:
MDL number:
UNSPSC Code:
12162002
assay
99%
refractive index
n20/D 1.516 (lit.)
density
1.164 g/mL at 25 °C (lit.)
SMILES string
CSCC(O)CO
Choose from one of the most recent versions:
Certificates of Analysis (COA)