Key Documents
216488
Supelco
Amberlite XAD4
20-60 mesh
Sign Into View Organizational & Contract Pricing
About This Item
CAS Number:
MDL number:
UNSPSC Code:
12000000
autoignition temp.
800 °F
matrix
styrene-divinylbenzene
particle size
20-60 mesh
pore size
~0.98 mL/g pore volume
100 Å mean pore size
density
1.02 g/mL (true wet)(lit.)
1.08 g/mL (skeletal)(lit.)
Looking for similar products? Visit Product Comparison Guide
General description
Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.
Application
Polyaromatic adsorbent for small hydrophobic compounds, surfactants, pharmaceutical manufacturing, phenols, chlorinated organics, pesticide removal and recovery, and organic removal from aqueous food streams.
Features and Benefits
polystyrene adsorbent having a high surface area
Other Notes
Two particle sizes are available:
Mesh 20-60 are Cat. Nos. 20276, 10358, XAD4
Mesh 20-50 is Cat. No. 06444
Legal Information
Amberlite is a trademark of DuPont de Nemours, Inc.
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)