Key Documents
207055
Supelco
Chromium(III) atomic absorption standard solution
1,000 μg/mL Cr(III) in 1 wt. % HCl (Exact Cr concentration on label)
Sign Into View Organizational & Contract Pricing
About This Item
UNSPSC Code:
41116107
PubChem Substance ID:
concentration
1,000 μg/mL Cr(III) in 1 wt. % HCl (Exact Cr concentration on label)
density
1.01 g/mL at 25 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)