203564
Lanthanum(III) sulfate nonahydrate
99.999% trace metals basis
About This Item
Recommended Products
assay
99.999% trace metals basis
form
powder
density
2.801 g/mL at 25 °C (lit.)
SMILES string
O.O.O.O.O.O.O.O.O.[La+3].[La+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?