Key Documents
198196
Sigma-Aldrich
Oil Red O
certified by the Biological Stain Commission, Dye content 85 %
Synonym(s):
1-([4-(Xylylazo)xylyl]azo)-2-naphthol, 1-[2,5-Dimethyl-4-(2,5-dimethylphenylazo)phenylazo]-2-naphthol, Solvent Red 27
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C26H24N4O
CAS Number:
Molecular Weight:
408.49
Colour Index Number:
26125
EC Number:
MDL number:
UNSPSC Code:
12171500
grade
certified by the Biological Stain Commission
composition
Dye content, 85%
mp
120 °C (dec.) (lit.)
SMILES string
Cc1ccc(C)c(c1)\N=N\c2cc(C)c(cc2C)\N=N/c3c(O)ccc4ccccc34
InChI
1S/C26H24N4O/c1-16-9-10-17(2)22(13-16)27-28-23-14-19(4)24(15-18(23)3)29-30-26-21-8-6-5-7-20(21)11-12-25(26)31/h5-15,31H,1-4H3
InChI key
NPGIHFRTRXVWOY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
biological stain
replaced by
Product No.
Description
Pricing
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)