Key Documents
178306
Sigma-Aldrich
N,S-Diacetylcysteamine
98%
Synonym(s):
N,S-Diacetyl-β-mercaptoethylamine, S-[2-(Acetyl­amino)ethyl] ethanethioate
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
CH3CONHCH2CH2SCOCH3
CAS Number:
Molecular Weight:
161.22
Beilstein/REAXYS Number:
1762551
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
98%
bp
181-182 °C/15 mmHg (lit.)
mp
29-31 °C (lit.)
SMILES string
CC(=O)NCCSC(C)=O
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)