Key Documents
170208
Sigma-Aldrich
Dipiperidinomethane
98%
Synonym(s):
1,1′-Methylenedipiperidine
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C11H22N2
CAS Number:
Molecular Weight:
182.31
EC Number:
MDL number:
UNSPSC Code:
12352100
Quality Level
assay
98%
refractive index
n20/D 1.482 (lit.)
bp
122-123 °C/15 mmHg (lit.)
density
0.915 g/mL at 25 °C (lit.)
SMILES string
C1CCN(CC1)CN2CCCCC2
pictograms
Exclamation mark
signalword
Warning
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
197.6 °F - closed cup
flash_point_c
92 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)