Key Documents
163309
Sigma-Aldrich
4,4′-Sulfonylbis(methyl benzoate)
technical grade, 90%
Synonym(s):
Dimethyl diphenyl sulfone 4,4′-dicarboxylate
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
O2S(C6H4CO2CH3)2
CAS Number:
Molecular Weight:
334.34
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade
technical grade
assay
90%
mp
195-196 °C (lit.)
SMILES string
COC(=O)c1ccc(cc1)S(=O)(=O)c2ccc(cc2)C(=O)OC
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.