Key Documents
150002
Sigma-Aldrich
Octaphenylcyclotetrasiloxane
97%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
[-Si(C6H5)2O-]4
CAS Number:
Molecular Weight:
793.17
Beilstein/REAXYS Number:
2320758
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
assay
97%
mp
196-198 °C (lit.)
SMILES string
O1[Si](O[Si](O[Si](O[Si]1(c2ccccc2)c3ccccc3)(c4ccccc4)c5ccccc5)(c6ccccc6)c7ccccc7)(c8ccccc8)c9ccccc9
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)