Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V000168

Sigma-Aldrich

L-Methionine

Vetec, 98.5%

Synonym(s):

(S)-2-Amino-4-(methylmercapto)butyric acid, L-2-Amino-4-(methylthio)butanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
149.21
Beilstein/REAXYS Number:
1722294
EC Number:
MDL number:
PubChem Substance ID:

product line

Vetec

assay

98.5%

mp

284 °C (dec.) (lit.)

SMILES string

CSCC[C@H](N)C(O)=O

InChI

1S/C5H11NO2S/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/t4-/m0/s1

InChI key

FFEARJCKVFRZRR-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irene Locatelli et al.
Hepatology (Baltimore, Md.), 60(2), 531-544 (2014-03-29)
Annexin A1 (AnxA1) is an effector of the resolution of inflammation and is highly effective in terminating acute inflammatory responses. However, its role in chronic settings is less investigated. Because changes in AnxA1 expression within adipose tissue characterize obesity in
Carmen E Perrone et al.
Experimental gerontology, 48(7), 654-660 (2012-07-24)
Restriction of dietary methionine by 80% slows the progression of aged-related diseases and prolongs lifespan in rodents. A salient feature of the methionine restriction phenotype is the significant reduction of adipose tissue mass, which is associated with improvement of insulin
Gad Galili et al.
Plant biotechnology journal, 11(2), 211-222 (2013-01-03)
Humans, as well as farm animals, cannot synthesize a number of essential amino acids, which are critical for their survival. Hence, these organisms must obtain these essential amino acids from their diets. Cereal and legume crops, which represent the major
Hwa-Young Kim
Antioxidants & redox signaling, 19(9), 958-969 (2012-12-04)
Selenium is utilized in the methionine sulfoxide reduction system that occurs in most organisms. Methionine sulfoxide reductases (Msrs), MsrA and MsrB, are the enzymes responsible for this system. Msrs repair oxidatively damaged proteins, protect against oxidative stress, and regulate protein
Ilayaraja Muthuramu et al.
Journal of molecular medicine (Berlin, Germany), 93(6), 609-618 (2015-04-11)
Plasma homocysteine levels predict heart failure incidence in prospective epidemiological studies. We evaluated whether selective homocysteine-lowering gene transfer beneficially affects cardiac remodeling and function in a model of pressure overload-induced cardiomyopathy induced by transverse aortic constriction (TAC). Female C57BL/6 low-density

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service