Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900793

Sigma-Aldrich

p-Toluic acid

Vetec, reagent grade, 98%

Synonym(s):

4-Methylbenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CO2H
CAS Number:
Molecular Weight:
136.15
Beilstein/REAXYS Number:
507600
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

bp

274-275 °C (lit.)

mp

177-180 °C (lit.)

SMILES string

Cc1ccc(cc1)C(O)=O

InChI

1S/C8H8O2/c1-6-2-4-7(5-3-6)8(9)10/h2-5H,1H3,(H,9,10)

Inchi Key

LPNBBFKOUUSUDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

357.8 °F

flash_point_c

181 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Balachandran et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 104, 114-129 (2012-12-26)
FT-Raman and FT-IR spectra for 2-hydroxy-p-toluic acid molecule had been recorded in the regions 3500-100 cm(-1) and 4000-400 cm(-1), respectively. Vibrational frequencies have been calculated in optimum state by employing density functional theory (DFT) and Hartree Fock (HF) methods with
Robert McGraw et al.
The Journal of chemical physics, 128(6), 064508-064508 (2008-02-20)
Recent kinetic extensions of the nucleation theorem suggest that the logarithm of the steady-state nucleation rate has strong multilinear dependence on the log concentrations of condensable species present in the vapor phase. A further remarkable result is that the coefficients
Qingjiang Wang et al.
Electrophoresis, 26(17), 3273-3278 (2005-09-07)
For its high reactivity and very short half-life, the hydroxyl radical (OH.) in vivo is very difficult to be detected. Usually, it is indirectly quantified by determining 2,3-dihydroxybenzoic acid (2,3-DHBA) and 2,5-dihydroxybenzoic acid (2,5-DHBA), which are the reaction products of
T Arras et al.
Journal of bacteriology, 180(8), 2133-2136 (1998-04-29)
The growth rates of Pseudomonas putida KT2442 and mt-2 on benzoate, 4-hydroxybenzoate, or 4-methylbenzoate showed an exponential decrease with decreasing oxygen tensions (partial O2 tension [pO2] values). The oxygen tensions resulting in half-maximal growth rates were in the range of
A C Franck-Mokross et al.
Applied microbiology and biotechnology, 50(2), 233-240 (1998-12-05)
Pseudomonas sp. D7-4 and Pseudomonas sp. B13 FR1(pFRC20P) degraded mixtures of chloro- and methyl-substituted benzoates exclusively via an extended ortho pathway, whereas in Pseudomonas putida WR 201 both ortho and meta fission were induced by mixtures of 3-chloro- and 3-methylbenzoate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service