Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900581

Sigma-Aldrich

2-Chlorobenzoic acid

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CO2H
CAS Number:
Molecular Weight:
156.57
Beilstein/REAXYS Number:
907340
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

mp

138-140 °C (lit.)

SMILES string

OC(=O)c1ccccc1Cl

InChI

1S/C7H5ClO2/c8-6-4-2-1-3-5(6)7(9)10/h1-4H,(H,9,10)

InChI key

IKCLCGXPQILATA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

343.4 °F - closed cup

flash_point_c

173 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A C Layton et al.
Journal of industrial microbiology, 13(6), 392-401 (1994-11-01)
The microbial populations in PCB-contaminated electric power substation capacitor bank soil (TVA soil) and from another PCB-contaminated site (New England soil) were compared to determine their potential to degrade PCB. Known biphenyl operon genes were used as gene probes in
B R Genthner
Biodegradation, 10(1), 27-33 (1999-07-29)
Dechlorination was the initial step of 2CB biodegradation in four 2-chlorobenzoate-degrading methanogenic consortia. Selected characteristics of ortho reductive dehalogenation were examined in consortia developed from the highest actively dechlorinating dilutions of the original 2CB consortia, designated consortia M34(-9), P20(-9), P21(-9)
Gang-hua Lang et al.
Applied microbiology and biotechnology, 83(6), 1085-1094 (2009-03-26)
TfdT is a LysR-type transcriptional regulator that activates the transcription of the chlorocatechol degradative gene operon tfdCDEF of the chlorobenzoate-degrading bacterium Burkholderia sp. NK8. To identify the amino acids involved in the effector recognition by TfdT, a polymerase-chain-reaction-based random mutagenesis
Grazia Baggi et al.
Biodegradation, 16(3), 275-282 (2005-05-04)
A study was made of the metabolic and co-metabolic intermediates of 2- and 3-chlorobenzoate, 2,3- and 3,5-dichlorobenzoate to elucidate the mechanism(s) involved in the negative effects observed on the growth of a chlorobenzoate-degrading microbial consortium in the presence of mixed
Meltem Urgun-Demirtas et al.
Water environment research : a research publication of the Water Environment Federation, 77(5), 511-518 (2005-11-09)
The feasibility of applying bacterial hemoglobin technology to degrade 2-chlorobenzoate (2-CBA) through co-metabolism under hypoxic conditions in a membrane bioreactor (MBR) process has been studied in the laboratory. 2-chlorobenzoate removal and chloride release rates in the MBR system varied from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service