Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900478

Sigma-Aldrich

ACES

Vetec, reagent grade, 98%

Synonym(s):

N-(2-Acetamido)-2-aminoethanesulfonic acid, N-(Carbamoylmethyl)-2-aminoethanesulfonic acid, N-(Carbamoylmethyl)taurine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCOCH2NHCH2CH2SO3H
CAS Number:
Molecular Weight:
182.20
Beilstein/REAXYS Number:
2253770
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

form

crystalline powder

pH

3.0-4.5

useful pH range

6.1-7.5

pKa (25 °C)

6.8

mp

>220 °C (dec.) (lit.)

solubility

water: 0.052 g/mL, clear, colorless

SMILES string

NC(=O)CNCCS(O)(=O)=O

InChI

1S/C4H10N2O4S/c5-4(7)3-6-1-2-11(8,9)10/h6H,1-3H2,(H2,5,7)(H,8,9,10)

InChI key

DBXNUXBLKRLWFA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher Oldfield et al.
Faraday discussions, 129, 247-263 (2005-02-18)
Water-in-oil microemulsions are thermodynamically stable single-phase dispersions of water and surfactant within a continuous oil phase. The classical ternary system, based on the surfactant sodium bis(2-ethylhexyl)sulfosuccinate ('AOT'), water and an alkane such as n-heptane, is an optically transparent monodispersion of
P H Edelstein et al.
Journal of clinical microbiology, 31(12), 3329-3330 (1993-12-01)
Growth of Legionella spp. on buffered charcoal yeast extract medium supplemented with alpha-ketoglutarate and formulated with 3-(n-morpholino)propanesulfonic acid (MOPS), 3-(n-morpholino)-2-hydroxypropanesulfonic acid (MOPSO), or n-(2-acetamido)-2-aminoethanesulfonic acid (ACES) buffer was similar. With three exceptions, growth was no different in buffered yeast extract
COMET challenges paradigm of ACE inhibition in mild heart failure.
Cardiovascular journal of South Africa : official journal for Southern Africa Cardiac Society [and] South African Society of Cardiac Practitioners, 14(4), 207-208 (2003-11-19)
D J Chappell
Clinical chemistry, 31(8), 1384-1386 (1985-08-01)
The hereditary deficiency variants of alpha 1-antitrypsin that are associated with diseases such as emphysema are usually identified by use of isoelectric focusing on polyacrylamide gels. Agarose is a simpler, faster, safer, and more reliable medium for this, but resolution
Free metal ion depletion by "Good's" buffers. II. N-(2-acetamido)-2-aminoethanesulfonic acid (ACESH): complexes with calcium(II), magnesium(II), manganese(II), cobalt(II), zinc(II), nickel(II), and copper(II).
J M Pope et al.
Analytical biochemistry, 103(2), 214-221 (1980-04-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service