Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900346

Sigma-Aldrich

DL-Isocitric acid trisodium salt hydrate

Vetec, reagent grade, 93%

Synonym(s):

threo-DsLs-Isocitric acid trisodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5Na3O7 · xH2O
CAS Number:
Molecular Weight:
258.07 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

93%

SMILES string

O.[Na+].[Na+].[Na+].OC(C(CC([O-])=O)C([O-])=O)C([O-])=O

InChI

1S/C6H8O7.3Na.H2O/c7-3(8)1-2(5(10)11)4(9)6(12)13;;;;/h2,4,9H,1H2,(H,7,8)(H,10,11)(H,12,13);;;;1H2/q;3*+1;/p-3

InChI key

BPEHKAXIRZOKIS-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ming-dong Wang et al.
Chinese medical journal, 124(17), 2611-2615 (2011-11-02)
Site A132Arg mutations potentially impair the affinity of isocitrate dehydrogenase 1 (IDH1) for its substrate isocitrate (ICT), consequently reducing the production of α-ketoglutarate and leading to tumor growth through the induction of the hypoxia-inducible factor-1 (HIF-1) pathway. However, given that
An-Ping Lin et al.
Biochemistry, 50(38), 8241-8250 (2011-08-25)
Yeast NAD(+)-specific isocitrate dehydrogenase is an allosterically regulated octameric enzyme composed of four heterodimers of a catalytic IDH2 subunit and a regulatory IDH1 subunit. Despite structural predictions that the enzyme would contain eight isocitrate binding sites, four NAD(+) binding sites
Metabolism, gliomas, and IDH1.
Jan Smeitink
The New England journal of medicine, 362(12), 1144-1145 (2010-03-26)
Zbigniew Lazar et al.
Bioresource technology, 102(13), 6982-6989 (2011-05-10)
Simultaneous production of citric acid (CA) and invertase by Yarrowia lipolytica A-101-B56-5 (SUC(+) clone) growing from sucrose, mixture of glucose and fructose, glucose or glycerol was investigated. Among the tested substrates the highest concentration of CA was reached from glycerol
Alexander B Taylor et al.
The Journal of biological chemistry, 283(16), 10872-10880 (2008-02-08)
Mitochondrial NAD(+)-specific isocitrate dehydrogenases (IDHs) are key regulators of flux through biosynthetic and oxidative pathways in response to cellular energy levels. Here we present the first structures of a eukaryotic member of this enzyme family, the allosteric, hetero-octameric, NAD(+)-specific IDH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service