Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900330

Sigma-Aldrich

Sodium propionate

Vetec, reagent grade, ≥99%

Synonym(s):

Propionic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2COONa
CAS Number:
Molecular Weight:
96.06
Beilstein/REAXYS Number:
3566934
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
grade:
reagent grade
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

≥99%

mp

285-286 °C (lit.)

SMILES string

[Na+].CCC([O-])=O

InChI

1S/C3H6O2.Na/c1-2-3(4)5;/h2H2,1H3,(H,4,5);/q;+1/p-1

InChI key

JXKPEJDQGNYQSM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katherine M Kennedy et al.
Journal of dairy science, 103(12), 11449-11460 (2020-11-24)
Our objective was to determine the temporal effects of increasing supply of propionate on propionate metabolism in liver tissue of dairy cows in the postpartum (PP) period. A total of 6 dairy cows [primiparous: n = 3, 9.00 ± 1.00
Yahong Cheng et al.
Biochemical pharmacology, 169, 113607-113607 (2019-09-07)
The present research was designed to evaluate the protective effects and underlying mechanisms of propionate, a bioactive food additive, on mitochondrial disruption, neuron necrosis and neurological deficits after epilepsy seizures. Epilepsy seizures was induced by repetitive injections of pentylenetetrazol at
Zhen Wang et al.
International immunopharmacology, 78, 106062-106062 (2019-12-13)
Alcoholic liver disease (ALD) presents one of the leading causes of cirrhosis worldwide. We have demonstrated that inulin alleviates ALD in mice. However, the exact role of hepatic macrophages in effects of inulin on ALD remains largely unclear. In vivo
M Duplessis et al.
Journal of dairy science, 100(10), 8578-8589 (2017-08-07)
This study was undertaken to evaluate the effect of supplementation of folic acid and vitamin B
Kelly A Foley et al.
Behavioural brain research, 278, 244-256 (2014-10-11)
Potential environmental risk factors for autism spectrum disorders (ASD) include viral/bacterial infection and an altered microbiome composition. The present study investigated whether administration of immune and gastrointestinal factors during gestation and early life altered startle response and prepulse inhibition in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service