Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PS858

Omite®

analytical standard

Synonym(s):

Propargite, 2-(4-tert-Butylphenoxy)cyclohexyl 2-propynyl sulfite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26O4S
CAS Number:
Molecular Weight:
350.47
Beilstein/REAXYS Number:
6934289
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 1000 mg

manufacturer/tradename

Chem Service, Inc. PS-858

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CC(C)(C)c1ccc(OC2CCCCC2OS(=O)OCC#C)cc1

InChI

1S/C19H26O4S/c1-5-14-21-24(20)23-18-9-7-6-8-17(18)22-16-12-10-15(11-13-16)19(2,3)4/h1,10-13,17-18H,6-9,14H2,2-4H3

InChI key

ZYHMJXZULPZUED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

Omite is a registered trademark of Chemtura Corporation

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

160.5 °F - Pensky-Martens closed cup

flash_point_c

71.4 °C - Pensky-Martens closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B K Kang et al.
Bulletin of environmental contamination and toxicology, 85(4), 414-418 (2010-08-11)
Residues of propargite were estimated in okra fruits by high performance liquid chromatography following single application of Omite 57 EC @570 and 1,140 g a.i./ha. Residues of propargite were confirmed by high performance thin layer chromatography. The average initial deposits
Mário Eidi Sato et al.
Experimental & applied acarology, 42(2), 107-120 (2007-06-06)
The purpose of this study was to evaluate the performance of Neoseiulus californicus (McGregor) for the control of Tetranychus urticae Koch in commercial strawberry fields, under greenhouse conditions, in association or not with the use of acaricides. The N. californicus
B K Kang et al.
Bulletin of environmental contamination and toxicology, 82(2), 248-250 (2008-11-15)
Residues of propargite were estimated in brinjal fruits by High Performance Liquid Chromatography (HPLC) following single application of Omite 57 EC @ 570 and 1140 g a.i./ha. The average initial deposits of propargite were observed to be 0.51 and 0.92
G A Herron et al.
Experimental & applied acarology, 29(1-2), 115-119 (2003-10-29)
Using susceptible and resistant reference strains and standard bioassay methods a new miticide product was screened for efficacy and cross-resistance against two-spotted spider mite, Tetranychus urticae Koch. Experimental product NN1 850.0 EW (fenpyroximate and propargite 3:10 ratio) was significantly (P<0.05)
Soumik Sarkar et al.
Journal of hazardous materials, 174(1-3), 295-298 (2009-11-07)
Biodegradation of miticide propargite was carried out in vitro by selected Pseudomonas strains isolated from tea rhizosphere. A total number of 13 strains were isolated and further screened based on their tolerance level to different concentrations of propargite. Five best

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service