Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PS654

Phorate

analytical standard

Synonym(s):

O,O-Diethyl S-(ethylthiomethyl) phosphorodithioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H17O2PS3
CAS Number:
Molecular Weight:
260.38
Beilstein/REAXYS Number:
1708517
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 100 mg

manufacturer/tradename

Chem Service, Inc. PS-654

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCOP(=S)(OCC)SCSCC

InChI

1S/C7H17O2PS3/c1-4-8-10(11,9-5-2)13-7-12-6-3/h4-7H2,1-3H3

InChI key

BULVZWIRKLYCBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monu Jariyal et al.
Ecotoxicology and environmental safety, 122, 24-30 (2015-07-18)
Use of phorate as a broad spectrum pesticide in agricultural crops is finding disfavor due to persistence of both the principal compound as well as its toxic residues in soil. Three phorate utilizing bacterial species (Pseudomonas sp. strain Imbl 4.3
Amal Chandra Das et al.
Chemosphere, 58(5), 579-584 (2004-12-29)
An experiment has been conducted under laboratory conditions to investigate the effect of phorate (an organophosphate insecticide) and carbofuran (a carbamate insecticide) at their recommended field rates (1.5 and 1.0 kga.i.ha-1, respectively) on the growth and multiplication of microorganisms as
Yuanyuan Zhang et al.
Ultrasonics sonochemistry, 19(1), 43-48 (2011-06-15)
The degradation of phorate in apple juice by sonication was investigated in the present study. Results showed that sonication was effective in eliminating phorate in apple juice, and the ultrasonic power and sonication time significantly influenced the degradation of phorate
D Ames Herbert et al.
Journal of economic entomology, 100(4), 1241-1247 (2007-09-14)
Tomato spotted wilt virus (family Bunyaviridae, genus Tospovirus, TSWV), transmitted by many thrips species, is a devastating pathogen of peanut, Arachis hypogaea L. TSWV has become a serious problem in the Virginia/Carolina peanut-growing region of the United States. During 2002
Rajeev Mahajan et al.
Environmental health perspectives, 114(8), 1205-1209 (2006-08-03)
We recently reported a link between use of the organophosphate pesticide phorate and risk of prostate cancer among applicators with a family history of prostate cancer in the Agricultural Health Study (AHS). This finding, together with findings of associations between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service