Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PS450

BDMC

analytical standard

Synonym(s):

4-Bromo-3,5-dimethylphenyl N-methylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NHCO2C6H2(CH3)2Br
CAS Number:
Molecular Weight:
258.11
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 100 mg

manufacturer/tradename

Chem Service, Inc. PS-450

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

114-116 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

CNC(=O)Oc1cc(C)c(Br)c(C)c1

InChI

1S/C10H12BrNO2/c1-6-4-8(14-10(13)12-3)5-7(2)9(6)11/h4-5H,1-3H3,(H,12,13)

InChI key

ZOZFMTULOYRWEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chien-Chih Yu et al.
In vivo (Athens, Greece), 29(6), 711-728 (2015-11-08)
Lung cancer is one of the most common malignancies and a predominant cause of cancer-related death. It can metastasize in almost all organs, and currently, while new cases are increasing, treatment is still insufficient. Bisdemethoxycurcumin (BDMC), one of the components
Jinhong Xu et al.
Molecular medicine reports, 12(5), 7603-7608 (2015-10-16)
Curcumin is an active component of the medicinal plant turmeric, which has been reported to have anti‑metastatic activities and induce autophagy in numerous cancer types. Bisdemethoxycurcumin (BDMC), one of the major active curcumin derivatives present in turmeric, was previously shown
Meyliana Wulandari et al.
Analytical and bioanalytical chemistry, 407(3), 803-812 (2014-07-25)
This paper describes the synthesis of novel molecularly imprinted polymers (MIPs), prepared by a noncovalent imprinting approach, for cleanup and preconcentration of curcumin (CUR) and bisdemethoxycurcumin (BDMC) from medicinal herbal extracts and further analysis by high-performance liquid chromatography with fluorescence

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service