Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PS262-250MG

Dexon

Synonym(s):

Fenaminosulf, 4-Dimethylaminobenzenediazosulfonic acid sodium salt, p-Diazodimethylaniline sodium sulfonate, Sodium [4-(dimethylamino)phenyl]diazenesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H10N3O3SNa
CAS Number:
Molecular Weight:
251.24
Beilstein/REAXYS Number:
4170376
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS262

SMILES string

[Na+].CN(C)c1ccc(cc1)N=NS([O-])(=O)=O

InChI

1S/C8H11N3O3S.Na/c1-11(2)8-5-3-7(4-6-8)9-10-15(12,13)14;/h3-6H,1-2H3,(H,12,13,14);/q;+1/p-1

InChI key

IWDQPCIQCXRBQP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Used in biological and pharmacological studies including:
  • Cloning and characterization of GABA transporter
  • Activity of anti-RSV carbonucleosides
  • Inhibition by fungicides of urea hydrolysis and nitrification of urea nitrogen in soil

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C B Howard et al.
The Journal of bone and joint surgery. British volume, 67(2), 313-316 (1985-03-01)
We have studied the use of filamentous polyglycolic acid (Dexon) for the induction of neotendon in the calcaneal tendon of sheep. Uniformly good results were obtained, with the formation of a thick neotendon. However, in a control group the power
F Bartone et al.
Urology, 29(5), 517-522 (1987-05-01)
In a study designed to determine the best suture for use in hypospadias surgery, polydioxanone (PDS), chromic catgut, and polyglycolic acid (Dexon) were studied under similar conditions in the penile foreskin of 16 baboons. Gross and microscopic observations were made
B Patsner et al.
Gynecologic oncology, 38(1), 146-148 (1990-07-01)
Three cases of postoperative enterocutaneous fistula formation following use of the Dexon mesh sling are reported from two gynecologic oncology services. Two patients had intestinal trauma or bowel resection at the time of mesh placement. Postoperative submesh abscess formation was
S Vainionpää et al.
Biomaterials, 8(1), 46-48 (1987-01-01)
The initial shear strength and changes in flexural strength of self-reinforced, absorbable polyglycolide (PGA) composite rods, submerged in distilled water (at 37 degrees C) for a period of 6 wk, were investigated. The recently developed self-reinforced absorbable material consists of
S B Pai
Mutation research, 119(3), 289-291 (1983-03-01)
Fenaminosulf (p-dimethylaminobenzenediazo sodium sulfonate, CAS registry No. 140-56-7) which is an active ingredient in several commercial fungicides was reported to be mutagenic in Salmonella typhimurium (McCann et al., 1975), Bacillus subtilis (Kada et al., 1974) and shown to cause chromosome

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service