Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PS2059

Fuberidazole

analytical standard

Synonym(s):

2-(2-Furanyl)-1H-benzimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8N2O
CAS Number:
Molecular Weight:
184.19
Beilstein/REAXYS Number:
153833
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 100 mg

manufacturer/tradename

Chem Service, Inc. PS-2059

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

c1ccc2[nH]c(nc2c1)-c3ccco3

InChI

1S/C11H8N2O/c1-2-5-9-8(4-1)12-11(13-9)10-6-3-7-14-10/h1-7H,(H,12,13)

InChI key

UYJUZNLFJAWNEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 Inhalation - Skin Sens. 1 - STOT RE 2

target_organs

Heart

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonia Moral et al.
Analytica chimica acta, 650(2), 207-213 (2009-09-02)
A supramolecular solvent consisting of vesicles, made up of equimolecular amounts of decanoic acid (DeA) and tetrabutylammonium decanoate (Bu4NDe), dispersed in a continuous aqueous phase, is proposed for the extraction of benzimidazolic fungicides (BFs) from fruits and vegetables. Carbendazim (CB)
A Garrido Frenich et al.
Analytical and bioanalytical chemistry, 375(7), 974-980 (2003-04-23)
The application of the generalised rank annihilation method (GRAM) and the trilinear decomposition (TLD) method to the resolution and quantitation of fluorescence excitation-emission matrices of a ternary mixture of pesticides, carbendazim, fuberidazole, and thiabendazole, with overlapped spectra is described. The
Gisela N Piccirilli et al.
The Analyst, 135(6), 1299-1308 (2010-04-17)
This paper presents a novel approach for the simultaneous determination of two widely used fungicides in a very interfering environment, combining the advantage of a spectrofluorimetric optosensor coupled to a flow-injection system and the selectivity of second-order chemometric algorithms. The
Adrian Frank et al.
Basic & clinical pharmacology & toxicology, 96(4), 325-330 (2005-03-10)
During metabolism of the fungicide 2-(2-furyl)benzimidazole (FB) in mammals, an optically active compound, metabolite A, was isolated. This compound, a principal metabolite in the biological degradation of FB, was isolated from the urine of horse and dog. When studied by
A Frank et al.
Toxicology letters, 17(3-4), 267-273 (1983-07-01)
The distribution of tritiated 2-(2-furyl)benzimidazole ([3H]FB) in male and pregnant albino mice and male pigmented mice was studied by whole-body autoradiography. The most notable finding was the accumulation of radioactivity in the melanin-containing tissues of the eye, the inner ear

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service