PS1096
Metasystox I
analytical standard
Synonym(s):
Demeton-S-methyl, Methyldemetone, S-(2-Ethylthioethyl) O,O-dimethyl phosphorothioate
About This Item
grade
analytical standard
packaging
ampule of 50 mg
manufacturer/tradename
Chem Service, Inc. PS-1096
application(s)
agriculture
environmental
storage temp.
−20°C
SMILES string
CCSCCSP(=O)(OC)OC
InChI
1S/C6H15O3PS2/c1-4-11-5-6-12-10(7,8-2)9-3/h4-6H2,1-3H3
InChI key
WEBQKRLKWNIYKK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Aquatic Chronic 2
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service