Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PS1063

Carbosulfan

analytical standard

Synonym(s):

2,3-Dihydro-2,2-dimethylbenzofuran-7-yl (dibutylaminothio)methylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32N2O3S
CAS Number:
Molecular Weight:
380.54
EC Number:
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

packaging

pkg of 250 mg

manufacturer/tradename

Chem Service, Inc. PS1063

concentration

250 mg/ampule neat

application(s)

agriculture
environmental

SMILES string

CCCCN(CCCC)SN(C)C(=O)Oc1cccc2CC(C)(C)Oc12

InChI

1S/C20H32N2O3S/c1-6-8-13-22(14-9-7-2)26-21(5)19(23)24-17-12-10-11-16-15-20(3,4)25-18(16)17/h10-12H,6-9,13-15H2,1-5H3

InChI key

JLQUFIHWVLZVTJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

204.8 °F - closed cup

flash_point_c

96 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Umetsu et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 15(1), 1-23 (1980-01-01)
The cleavage of the N-S bond in 2,3-dihydro-2,2-dimethyl-7-benzofuranyl (di-n-butylaminosulfenyl) (methyl)carbamate was examined in different buffer solutions (hydrolysis), in buffer solution containing sulfhydryl reagents (thiolysis) and on thin-layer chromatographic plates. In buffer solution and on thin-layer plates, N-S bond cleavage readily

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service