Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PS1003

Isofenphos

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24NO4PS
CAS Number:
Molecular Weight:
345.39
Beilstein/REAXYS Number:
2949615
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

pkg of 100 mg

manufacturer/tradename

Chem Service, Inc. PS-1003

application(s)

agriculture
environmental

SMILES string

CCOP(=S)(NC(C)C)Oc1ccccc1C(=O)OC(C)C

InChI

1S/C15H24NO4PS/c1-6-18-21(22,16-11(2)3)20-14-10-8-7-9-13(14)15(17)19-12(4)5/h7-12H,6H2,1-5H3,(H,16,22)

InChI key

HOQADATXFBOEGG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Tracey et al.
Human & experimental toxicology, 9(2), 99-100 (1990-03-01)
Two cases of organophosphorus poisoning are reported. Both were treated with a combination of atropine and glycopyrrolate as well as benzodiazepine and pralidoxime. The advantages of glycopyrrolate over atropine are discussed.
M A Rodrigues et al.
Biomedical and environmental sciences : BES, 8(1), 18-22 (1995-03-01)
The cytotoxicity of isophenphos, an organophosphorus insecticide that has the potential to cause delayed polyneuropathy, was evaluated in GBK and V79 cells. A 72 h time course following isophenphos exposure indicated a dose-dependent growth inhibition as determined by cell counts.
Degradation of diazinon, chlorpyrifos, isofenphos, and pendimethalin in grass and compost.
C R Lemmon et al.
Bulletin of environmental contamination and toxicology, 48(3), 409-415 (1992-03-11)
E E Calore et al.
Ecotoxicology and environmental safety, 43(2), 187-194 (1999-06-22)
The degenerative process of the myofibers of the diaphragm of rats intoxicated with the organophosphate isofenphos, a compound that inhibits esterases, was studied at different intervals of intoxication. Early disorganization of the intermyofibrillar network and of the myofilaments, as well
M J Cavaliere et al.
Journal of toxicology. Clinical toxicology, 36(4), 295-300 (1998-08-26)
To determine the protective effect of pralidoxime on muscle fiber necrosis induced by organophosphate acute intoxication in rats. Adult male Wistar rats were given oral organophosphate compounds dissolved in glycerol formal: dichlorvos, isofenphos, metamidophos, and diazinon. Half of the animals

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service