Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MET678B

Supelco

3-Methyl-4-nitrophenol

analytical standard

Synonym(s):

4-Nitro-m-cresol, 5-Hydroxy-2-nitrotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)OH
CAS Number:
Molecular Weight:
153.14
Beilstein/REAXYS Number:
1868105
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 1000 mg

manufacturer/tradename

Chem Service, Inc. MET-678B

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

125-130 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Cc1cc(O)ccc1[N+]([O-])=O

InChI

1S/C7H7NO3/c1-5-4-6(9)2-3-7(5)8(10)11/h2-4,9H,1H3

InChI key

PIIZYNQECPTVEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuling Mi et al.
Toxicology letters, 190(1), 61-65 (2009-07-15)
Diesel exhaust particles (DEP) are considered to be one of the most important air pollutants. In this study, the protective effect of quercetin, an antioxidant flavonoid, on oxidative damage of testicular cells was studied by analysis of the intracellular antioxidant
Zhuo Yue et al.
Environmental toxicology, 27(11), 653-661 (2011-03-05)
The 3-methyl-4-nitrophenol (4-nitro-m-cresol; PNMC) exists in diesel exhaust particles (DEP), and is also one of the degradation products of insecticide fenitrothion. To assess potential nephrotoxicity of PNMC, male Sprague-Dawley (SD) rats were subcutaneously dosed with PNMC at 1, 10, and
Chunmei Li et al.
Biological & pharmaceutical bulletin, 30(12), 2376-2380 (2007-12-07)
In previous studies, we found that 3-methyl-4-nitrophenol (4-nitro-m-cresol; PNMC) isolated from diesel exhaust particles, and also a degradation product of the insecticide fenitrothion, exhibited testicular toxicity in the male of both immature rat and adult Japanese quail. It is well
ChunMei Li et al.
European journal of pharmacology, 543(1-3), 194-199 (2006-07-11)
In our continuing studies on nitrophenol derivatives as vasodilators in diesel exhaust particles, we have reported that nitrophenols in diesel exhaust particles possess not only vasodilatory activity but also estrogenic activity in vitro and in vivo, as well as anti-androgenic
Qing Li et al.
Human & experimental toxicology, 30(7), 665-674 (2010-07-14)
Fenitrothion (FNT) is used throughout the world as an insecticide in agriculture. To investigate the effect of FNT on the splenocytes and the underlying mechanism, FNT and its main metabolite, 3-methyl-4-nitrophenol (MNP), were administered orally to Wistar rats in daily

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service