Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48695

Supelco

4-Nitrophenol solution

500 μg/mL in methanol, analytical standard

Synonym(s):

p-Nitrophenol solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO3
CAS Number:
Molecular Weight:
139.11
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

CofA

certificate of analysis is enclosed in each package.

packaging

ampule of 1 mL

concentration

500 μg/mL in methanol

application(s)

environmental

SMILES string

Oc1ccc(cc1)[N+]([O-])=O

InChI

1S/C6H5NO3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8H

InChI key

BTJIUGUIPKRLHP-UHFFFAOYSA-N

Gene Information

human ... UGT1A4(54657)

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

51.8 °F - closed cup

flash_point_c

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tang Qiong et al.
Applied biochemistry and biotechnology, 169(7), 2126-2137 (2013-02-13)
Effects of 4-nitrophenol (4NP) on the overall performance of membrane bioreactors (MBRs) were investigated in two bench-scale submerged MBRs. Positive impacts of 4NP on activated sludge production and membrane fouling were demonstrated over 45 days of stable operational period. After
Xiaomei Wang et al.
Journal of nanoscience and nanotechnology, 13(3), 2218-2223 (2013-06-13)
Adsorption equilibria and kinetics of p-nitrophenol adsorbed on HJ-01 were investigated in this study. The result indicated that phenoxy groups were uploaded on the skeleton of HJ-01 successfully, the Friedel-Crafts reaction brought on prodigious changes for the Brunauer-Emmett-Teller (BET) specific
Ali Mehdinia et al.
Journal of chromatography. A, 1283, 82-88 (2013-03-08)
In this study, an analytical procedure for the selective extraction and detection of 4-nitrophenol (4-NP) was investigated by using of molecularly imprinted polymer on the surface of magnetic nanoparticles (MNPs). The magnetic nanoparticles were modified by tetraethyl orthosilicate (TEOS) and
Jau-Rung Chiou et al.
Journal of hazardous materials, 248-249, 394-400 (2013-02-19)
Silver/iron oxide composite nanoparticles have been synthesized successfully via a facile one-pot green route by the use of l-arginine, which created an aqueous solution of about pH 10 and acted as a reducing agent for the successive formation of iron
Simon De Corte et al.
Colloids and surfaces. B, Biointerfaces, 102, 898-904 (2012-10-31)
An increasing demand for catalytic Pd nanoparticles has motivated the search for sustainable production methods. An innovative approach uses bacterial cells as support material for synthesizing Pd nanoparticles by reduction of Pd(II) with e.g. hydrogen or formate. Nevertheless, drawbacks of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service