Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48539

Supelco

Bromoform

analytical standard, ampule of 5000 mg

Synonym(s):

Tribromomethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHBr3
CAS Number:
Molecular Weight:
252.73
Beilstein/REAXYS Number:
1731048
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor density

8.7 (vs air)

vapor pressure

5 mmHg ( 20 °C)

packaging

ampule of 5000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.595 (lit.)

bp

146-150 °C (lit.)

mp

5-8 °C (lit.)

density

2.89 g/mL at 25 °C (lit.)

application(s)

environmental

storage temp.

2-8°C

SMILES string

BrC(Br)Br

InChI

1S/CHBr3/c2-1(3)4/h1H

InChI key

DIKBFYAXUHHXCS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

106.3 °F - closed cup - (own results)

flash_point_c

41.3 °C - closed cup - (own results)

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bichismita Sahu et al.
The Journal of organic chemistry, 74(6), 2601-2604 (2009-02-26)
Addition of bromoform to conjugated nitroalkenes in the presence of Mg provided beta-tribromomethyl nitroalkanes in good to excellent yields and diastereoselectivity. These novel Michael adducts, formed under radical conditions, underwent elimination of HBr in the same pot under reflux to
M Valcke et al.
Regulatory toxicology and pharmacology : RTP, 59(2), 258-269 (2010-10-26)
The objective of this study was to assess the impact of the exposure route on the human kinetic adjustment factor (HKAF), for which a default value of 3.16 is used in non-cancer risk assessment. A multi-route PBPK model was modified
Lisa George et al.
The Journal of chemical physics, 135(12), 124503-124503 (2011-10-07)
Iso-polyhalomethanes are known reactive intermediates that play a pivotal role in the photochemistry of halomethanes in condensed phases. In this work, iso-bromoform (iso-CHBr(3)) and its deuterated isotopomer were characterized by matrix isolation infrared and UV/visible spectroscopy, supported by ab initio
Szilvia Pothoczki et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 22(40), 404211-404211 (2011-03-10)
New neutron and x-ray diffraction measurements are reported on liquid chloroform, CHCl(3), and bromoform, CHBr(3). Experimental total scattering structure factors have been interpreted by the reverse Monte Carlo method of structural modelling. Partial radial distribution functions, intramolecular bond angle distributions
Carolina Lourencetti et al.
Environment international, 45, 59-67 (2012-05-11)
This first study of trihalomethanes (THMs) in swimming pools using bromine agents for water disinfection under real conditions shows that the mixtures of these compounds are largely dominated by bromoform in a similar process as chloroform becomes the dominant THM

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service