Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48514

Supelco

1,1,2,2-Tetrachloroethane

analytical standard, ampule of 1000 mg

Synonym(s):

Acetylene tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CHCl2CHCl2
CAS Number:
Molecular Weight:
167.85
Beilstein/REAXYS Number:
969206
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor density

5.8 (vs air)

vapor pressure

8 mmHg ( 20 °C)

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.494 (lit.)

bp

147 °C (lit.)

mp

−43 °C (lit.)

density

1.586 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

SMILES string

ClC(Cl)C(Cl)Cl

InChI

1S/C2H2Cl4/c3-1(4)2(5)6/h1-2H

InChI key

QPFMBZIOSGYJDE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

recommended

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Aquatic Chronic 2

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wan-Chun Tsai et al.
Journal of chromatography. A, 1216(45), 7846-7850 (2009-10-07)
A novel sample preparation method "Dispersive liquid-liquid-liquid microextraction" (DLLLME) was developed in this study. DLLLME was combined with liquid chromatography system to determine chlorophenoxy acid herbicide in aqueous samples. DLLLME is a rapid and environmentally friendly sample pretreatment method. In
Marcos Caroli Rezende et al.
Magnetic resonance in chemistry : MRC, 47(6), 505-510 (2009-04-01)
Spectra of the N-phenyl-5,6-dihydro-2,4-diphenylbenzo[h]quinolinium tetrafluoroborate (1) and of the N-phenyl-5,6,8,9-tetrahydro-7-phenyldibenzo[c,h]acridinium tetrafluoroborate (2) were recorded in various solvents and temperatures. The analysis of the (1)H-NMR spectra of the tetrafluoroborate salt 1, recorded in acetone, acetonitrile, 1,1,2,2-tetrachloroethane and chloroform, revealed the existence
Srigokul Upadhyayula et al.
The journal of physical chemistry. B, 115(30), 9473-9490 (2011-06-21)
Electrostatic properties of proteins are crucial for their functionality. Carboxyamides are small polar groups that, as peptide bonds, are principal structural components of proteins that govern their electrostatic properties. We investigated the medium dependence of the molar polarization and of
Debolina Basu et al.
Bioresource technology, 101(1), 21-25 (2009-08-25)
This study examines the performance of bench-scale Upflow Anaerobic Sludge Blanket (UASB) reactor in treating simulated wastewaters containing 1,1,2,2-tetrachloroethane (TeCA). Reactors R1 (control) and R2 (containing TeCA) were operated at Hydraulic Retention Times (HRTs) of 36, 30, 24, 18 and
Martin Elsner et al.
Environmental science & technology, 41(11), 4111-4117 (2007-07-07)
Despite widespread implementation of zero-valent iron remediation schemes, the manner and order of chemical bond cleavage in iron-mediated organohalide transformations remains imperfectly understood. We present insights from carbon isotope fractionation for the dehalogenation of 1,1,2,2-tetrachloroethane (1,1,2,2-TeCA) and 1,1,1-trichloroethane (1,1,1-TCA) by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service