Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48370

Supelco

Chlorobenzilate solution

200 μg/mL in methylene chloride, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

CofA

certificate of analysis is enclosed in each package.

packaging

ampule of 1 mL

concentration

200 μg/mL in methylene chloride

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

SMILES string

CCOC(=O)C(O)(c1ccc(Cl)cc1)c2ccc(Cl)cc2

InChI

1S/C16H14Cl2O3/c1-2-21-15(19)16(20,11-3-7-13(17)8-4-11)12-5-9-14(18)10-6-12/h3-10,20H,2H2,1H3

InChI key

RAPBNVDSDCTNRC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Exposure of spray applicators and mixer-loaders to chlorobenzilate miticide in Florida citrus groves.
H N Nigg et al.
Archives of environmental contamination and toxicology, 12(4), 477-482 (1983-07-01)
Y S Wang et al.
Ecotoxicology and environmental safety, 28(2), 193-200 (1994-07-01)
The dissipation of acaricide chlorobenzilate in selected soil and river water samples was studied. Under incubation conditions at 25 degrees C and 90% field capacity of moisture for 21 days, the residues of chlorobenzilate were 14 and 55% in Lukang
Prediction of pesticide dermal exposure and urinary metabolite level of tree crop harvesters from field residues.
J H Stamper et al.
Bulletin of environmental contamination and toxicology, 36(5), 693-700 (1986-05-01)
S Flodström et al.
Cancer letters, 43(3), 161-166 (1988-12-15)
The mouse liver carcinogen chlorobenzilate (CB), a DDT-related pesticide, was investigated for enhancement of enzyme altered foci incidence in partially hepatectomized, diethyl-nitrosamine-initiated rats. In this in vivo experiment, CB administered per os (25 or 100 mg/kg per day for 10
Effects of certain pesticides on the embryogenesis of Boophilus annulatus (Say). Ixodoidea. Ixodidae.
K M el-Kammah et al.
Journal of the Egyptian Society of Parasitology, 17(2), 583-589 (1987-12-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service