Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47949

Supelco

Ethylene oxide solution

2000 μg/mL in methylene chloride, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4O
CAS Number:
Molecular Weight:
44.05
MDL number:
UNSPSC Code:
12000000

grade

analytical standard

CofA

certificate of analysis is enclosed in each package.

feature

standard type calibration

packaging

ampule of 1 mL

concentration

2000 μg/mL in methylene chloride

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

C1CO1

InChI

1S/C2H4O/c1-2-3-1/h1-2H2

Inchi Key

IAYPIBMASNFSPL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Muta. 1B - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

212.0 °F

flash_point_c

100 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthieu Gatineau et al.
American journal of veterinary research, 73(10), 1665-1669 (2012-09-28)
To compare effects of sterilization with hydrogen peroxide gas plasma (HPGP), ethylene oxide, and steam on bioadhesive properties of nylon and polyethylene lines used for stabilization of canine stifle joints. Samples of a 36.3-kg test nylon leader line, 57.8-kg test
Filippo Doria et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(1), 78-81 (2012-12-06)
Caught in the oxirane: Naphthalene diimides conjugated to a quinone methide and an oxirane have been synthesized and investigated as selective DNA G-quadruplex alkylating agents. The oxirane derivative generates a stable adduct with a G-quadruplex and shows selective alkylation of
A T Natarajan et al.
Mutation research, 330(1-2), 55-70 (1995-08-01)
A risk estimate of the heritable effects of ethylene oxide exposure, using the parallelogram approach, as suggested by Frits Sobels, is described. The approach is based on available data on the ethylene oxide-induced responses for the same genetic endpoint in
L Ehrenberg et al.
Mutation research, 330(1-2), 41-54 (1995-08-01)
Data for relationships between in vivo doses inferred from levels of hemoglobin (Hb) or DNA adducts and administered (by inhalation or injection) doses of ethylene oxide (EO) in mice, rats and humans are reviewed. At low absorbed doses or dose
R Thier et al.
Critical reviews in toxicology, 30(5), 595-608 (2000-10-31)
Long-term inhalation studies in rodents have presented unequivocal evidence of experimental carcinogenicity of ethylene oxide, based on the formation of malignant tumors at multiple sites. However, despite a considerable body of epidemiological data only limited evidence has been obtained of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service