Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47877

Supelco

Maltotetraose, DP4

analytical standard

Synonym(s):

Maltotetraose, Amylotetraose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H42O21
CAS Number:
Molecular Weight:
666.58
Beilstein/REAXYS Number:
78870
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:

grade

analytical standard

feature

standard type oligosaccharide

analyte chemical class(es)

oligosaccharides

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@@H]2CO)O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]4CO)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C24H42O21/c25-1-5-9(29)10(30)15(35)22(40-5)44-19-7(3-27)42-24(17(37)12(19)32)45-20-8(4-28)41-23(16(36)13(20)33)43-18-6(2-26)39-21(38)14(34)11(18)31/h5-38H,1-4H2/t5-,6-,7-,8-,9-,10+,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21+,22-,23-,24-/m1/s1

InChI key

LUEWUZLMQUOBSB-ZLBHSGTGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rupendra Mukerjea et al.
Carbohydrate research, 338(18), 1811-1822 (2003-08-23)
D-[14C]glucose was incorporated into starch when 12 varieties of starch granules were incubated with [14C]sucrose. Digestion of the 14C-labeled starches with porcine pancreatic alpha amylase showed that a high percentage (16.1-84.1%) of the synthesized starch gave a relatively high molecular
Heidi Neugebauer et al.
Journal of bacteriology, 187(24), 8300-8311 (2005-12-03)
Analysis of the genome sequence of Caulobacter crescentus predicts 67 TonB-dependent outer membrane proteins. To demonstrate that among them are proteins that transport nutrients other than chelated Fe(3+) and vitamin B(12)-the substrates hitherto known to be transported by TonB-dependent transporters-the
P Degn et al.
Carbohydrate research, 329(1), 57-63 (2000-11-22)
Glucose and maltose esters were synthesised in organic media by employing a lipase (E.C. 3.1.1.3) from Candida antarctica. In a second reaction step, a transglycosylation catalysed by a cyclodextrin glycosyltransferase (E.C. 2.4.1.19) from either Paenibacillus sp. F8 or Bacillus sp.
A W de Boer et al.
Pediatric nephrology (Berlin, Germany), 15(1-2), 21-24 (2000-11-30)
Icodextrin use in adults provides sustained ultrafiltration (UF) in long-term dwells. No information is available on UF and metabolism in children. In 11 children, a volume of 1,049+/-138 ml/m2 of the study fluid (1.36% glucose, 7.5% icodextrin, 3.86% glucose) was
Wim Sluiter et al.
Clinical chemistry, 58(7), 1139-1147 (2012-05-25)
Urinary excretion of the tetrasaccharide 6-α-D-glucopyranosyl-maltotriose (Glc₄) is increased in various clinical conditions associated with increased turnover or storage of glycogen, making Glc₄ a potential biomarker for glycogen storage diseases (GSD). We developed an ultraperformance liquid chromatography-tandem mass spectrometry (UPLC-MS/MS)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service