Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47752-U

Supelco

2,2-Dichloropropane

analytical standard

Synonym(s):

Isopropylidene dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CCl2CH3
CAS Number:
Molecular Weight:
112.99
Beilstein/REAXYS Number:
1731150
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.415 (lit.)

bp

68-69 °C (lit.)

mp

−35 °C (lit.)

density

1.082 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

room temp

SMILES string

CC(C)(Cl)Cl

InChI

1S/C3H6Cl2/c1-3(2,4)5/h1-2H3

InChI key

ZEOVXNVKXIPWMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

23.0 °F - closed cup

flash_point_c

-5.0 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service