Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442764

Supelco

Pronamide (Kerb)

analytical standard

Synonym(s):

Propyzamide, Pronamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11Cl2NO
CAS Number:
Molecular Weight:
256.13
Beilstein/REAXYS Number:
882391
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-30°C

SMILES string

CC(C)(NC(=O)c1cc(Cl)cc(Cl)c1)C#C

InChI

1S/C12H11Cl2NO/c1-4-12(2,3)15-11(16)8-5-9(13)7-10(14)6-8/h1,5-7H,2-3H3,(H,15,16)

InChI key

PHNUZKMIPFFYSO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Health hazardEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K R Häntzschel et al.
Protoplasma, 241(1-4), 99-104 (2010-02-04)
Mitosis in plants can be blocked by colchicine which has the capacity to bind microtubule subunits. In maize (Zea mays L.) breeding, it is frequently being used for doubling chromosome numbers of haploids for producing homozygous doubled haploids. However, colchicine
K Yoshida et al.
Protoplasma, 221(3-4), 227-235 (2003-06-13)
Some species of Spirogyra form rosette-shaped or rod-shaped rhizoids in the terminal cell of the filaments. In the present study, we analyzed an involvement of microtubules (MTs) in rhizoid differentiation. Before rhizoid differentiation, cortical MTs were arranged transversely to the
Maria Kwiatkowska et al.
Cell biology international, 36(5), 455-462 (2012-02-03)
Lipid bodies present in lipotubuloids of Ornithogalum umbellatum ovary epidermis take the form of a lens between leaflets of ER (endoplasmic reticulum) membrane filled with a highly osmiophilic substance. The two enzymes, DGAT1 [DAG (diacylglycerol) acyltransferase 1] and DGAT2 (DAG
A Tediosi et al.
The Science of the total environment, 438, 103-112 (2012-09-18)
Propyzamide and carbetamide are essential for blackgrass control in oilseed rape production. However, both of these compounds can contaminate surface waters and pose compliance problems for water utilities. The transport of propyzamide and carbetamide to an instrumented field drain in
Aaron A Santner et al.
The Plant journal : for cell and molecular biology, 45(5), 752-764 (2006-02-08)
The WAG1 and WAG2 genes of Arabidopsis thaliana encode protein-serine/threonine kinases that are closely related to PINOID. In order to determine what roles WAG1 and WAG2 play in seedling development, we used a reverse genetics approach to study the wag1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service