Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442590

Supelco

Etridiazole

PESTANAL®, analytical standard

Synonym(s):

5-Ethoxy-3-trichloromethyl-1,2,4-thiadiazole, Ethazol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5Cl3N2OS
CAS Number:
Molecular Weight:
247.53
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

packaging

ampule of 250 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

room temp

SMILES string

CCOc1nc(ns1)C(Cl)(Cl)Cl

InChI

1S/C5H5Cl3N2OS/c1-2-11-4-9-3(10-12-4)5(6,7)8/h2H2,1H3

InChI key

KQTVWCSONPJJPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Etridiazole is an agricultural fungicide,[1] which is found to be effective against fungi of the oomycete group.[2] Its mode of action involves the activation of phospholipases in the mitochondrial membranes.[2]

application

Etridiazole may be used as a reference standard to examine the pesticide residues in fruits[3], cereals[4] and edible vegetable oils[5] by using routine gas chromatographic method.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

310.1 °F - open cup

flash_point_c

154.5 °C - open cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A rapid microbioassay for discovery of novel fungicides for Phytophthora spp.
Kuhajek J M, et al.
Phytopathology, 93(1), 46-53 (2003)
Determination of 255 pesticides in edible vegetable oils using QuEChERS method and gas chromatography tandem mass spectrometry.
He Z, et al.
Analytical and Bioanalytical Chemistry, 409(4), 1017-1030 (2017)
P P Golikov et al.
Voprosy meditsinskoi khimii, 40(2), 9-12 (1994-03-01)
The effect of sodium sulfaethidole on Types II and III glucocorticoid receptors was studied in liver cytosol fraction obtained from female Wistar rats of 180-200 g body weight. Functions of Type II glucocorticoid receptors in liver cytosol were evaluated by
[Severe manifestations of drug allergy in children].
T N Sukovatykh et al.
Pediatriia, (5)(5), 76-77 (1982-05-01)
On the mode of action of the fungicide etridiazole.
Radzuhn B and Lyr H
Pesticide Biochemistry and Physiology, 22(1), 14-23 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service